siRNA Details
Gene Symbol: Slc6a4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010484.2 | NP_034614.2 | 9 | 1407 | ||
GenBank mRNA | AF013604.1 | - | 8 | 1220 | ||
AK078634.1 | - | 1 | 964 | |||
BC108978.1 | - | 9 | 1389 | 40047534 | ||
BC108979.1 | - | 9 | 1389 | 40047537 | ||
X66119.1 | - | 8 | 1001 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
