siRNA Details
Gene Symbol: Elavl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010485.3 | NP_034615.2 | 2, 3 | 397 | ||
GenBank mRNA | AK034536.1 | - | 2, 3 | 384 | ||
AK075818.1 | - | 2, 3 | 359 | |||
AK078333.1 | - | 2, 3 | 713 | |||
AK080365.1 | - | 2, 3 | 355 | |||
AK089088.1 | - | 1, 2 | 104 | |||
AK148528.1 | - | 2, 3 | 325 | |||
AK161667.1 | - | 1, 2 | 114 | |||
AK180399.1 | - | 1, 2 | 138 | |||
AK180790.1 | - | 1, 2 | 138 | |||
AK190746.1 | - | 1, 2 | 138 | |||
AK197032.1 | - | 1, 2 | 138 | |||
AK198784.1 | - | 1, 2 | 89 | |||
AK203188.1 | - | 1, 2 | 138 | |||
U65735.1 | - | 1, 2 | 166 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
