siRNA Details
Gene Symbol: Elavl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010487.2 | NP_034617.1 | 2 | 633 | ||
GenBank mRNA | AK090281.1 | - | 2 | 633 | ||
BC052097.1 | - | 2 | 620 | 5695543 | ||
U29148.1 | - | 2 | 612 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
