siRNA Details
Gene Symbol: Elavl4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001038698.1 | NP_001033787.1 | 3 | 542 | ||
NM_001163397.1 | NP_001156869.1 | 3 | 378 | |||
NM_001163399.1 | NP_001156871.1 | 3 | 628 | |||
NM_010488.4 | NP_034618.2 | 3 | 628 | |||
GenBank mRNA | AF041341.1 | - | 3 | 390 | ||
AK014133.1 | - | 3 | 396 | |||
AK079088.1 | - | 3 | 533 | |||
BB648788.1 | - | 3 | 629 | |||
BC048159.1 | - | 3 | 601 | 5703005 | ||
BC051987.1 | - | 299 | ||||
BC052451.1 | - | 3 | 505 | 6409658 | ||
CF726962.1 | - | 3 | 378 | |||
CN663544.1 | - | 3 | 542 | |||
D31953.1 | - | 3 | 375 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001077651, XM_006238593, XM_006238592, XM_006238591, XM_006238595, and others.
Gene Ontology Categories:
