siRNA Details
Gene Symbol: Idua | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008325.4 | NP_032351.2 | 5, 6 | 912 | ||
NM_172997.1 | NP_766585.1 | 4, 5 | 771 | |||
GenBank mRNA | AK031414.1 | - | 4, 5 | 566 | ||
AK045330.1 | - | 3, 4 | 399 | |||
AK155627.1 | - | 5, 6 | 913 | |||
BC057935.1 | - | 4, 5 | 533 | 5327150 | ||
L34111.1 | - | 4, 5 | 486 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
