siRNA Details
Gene Symbol: Ier2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010499.4 | NP_034629.3 | 1 | 936 | ||
GenBank mRNA | AK032692.1 | - | 934 | |||
AK144601.1 | - | 1 | 933 | |||
AK150860.1 | - | 1 | 931 | |||
AK152747.1 | - | 1 | 931 | |||
AK170433.1 | - | 933 | ||||
AK172332.1 | - | 1 | 879 | |||
BC002067.1 | - | 1 | 921 | 3590296 | ||
M31042.1 | - | 1 | 935 | |||
M59821.1 | - | 1 | 938 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
