siRNA Details
Gene Symbol: Ifna4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC116217.2 | - | 1 | 310 | 40091191 | |
BC119349.1 | - | 1 | 355 | 8734098 | ||
BC119351.1 | - | 1 | 355 | 8734100 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
