siRNA Details
Gene Symbol: Ifna6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_206871.1 | NP_996754.1 | 1 | 123 | ||
GenBank mRNA | BC117774.1 | - | 123 | 40091177 |
Gene Symbol: Ifna1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010502.2 | NP_034632.2 | 1 | 123 |
Gene Symbol: Ifnab | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008336.2 | NP_032362.2 | 1 | 164 | ||
GenBank mRNA | BC116862.1 | - | 1 | 164 | 40126181 | |
BC116864.1 | - | 1 | 164 | 40126183 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
