siRNA Details
Gene Symbol: Ifna7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008334.3 | NP_032360.2 | 1 | 121 | ||
GenBank mRNA | BC130231.1 | - | 1 | 122 | 40141930 |
Gene Symbol: Ifna5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010505.2 | NP_034635.2 | 1 | 118 | ||
GenBank mRNA | BC120910.1 | - | 1 | 118 | 40091152 | |
BC120911.1 | - | 1 | 118 | 40091153 |
Gene Symbol: Ifna6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC151087.1 | - | 1 | 258 | 9088000 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
