siRNA Details
Gene Symbol: Ifnar2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010509.2 | NP_034639.2 | 7, 8 | 1047 | ||
GenBank mRNA | AF013274.1 | - | 7, 8 | 825 | ||
AK139169.1 | - | 7, 8 | 1027 | |||
AK150386.1 | - | 7, 8 | 981 | |||
AK150597.1 | - | 7, 8 | 960 | |||
AK151474.1 | - | 7, 8 | 975 | |||
AK151493.1 | - | 7, 8 | 960 | |||
AK151777.1 | - | 7, 8 | 975 | |||
AK152878.1 | - | 7, 8 | 981 | |||
AK153317.1 | - | 7, 8 | 981 | |||
BC071225.1 | - | 6, 7 | 745 | 4187603 | ||
Y09865.1 | - | 7, 8 | 937 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
