siRNA Details
Gene Symbol: Ifrd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013562.2 | NP_038590.2 | 7 | 974 | ||
GenBank mRNA | AK078888.1 | - | 972 | |||
AK168682.1 | - | 7 | 971 | |||
AK215125.1 | - | 3 | 318 | |||
AK219989.1 | - | 3 | 200 | |||
BC037013.1 | - | 7 | 948 | |||
BC043723.1 | - | 7 | 941 | 5369100 | ||
X17400.1 | - | 7 | 951 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
