siRNA Details
Gene Symbol: Cd79b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008339.2 | NP_032365.1 | 3 | 351 | ||
GenBank mRNA | AK143573.1 | - | 3 | 352 | ||
BC012226.1 | - | 3 | 351 | 4164551 | ||
CT010358.1 | - | 3 | 306 | |||
J03857.1 | - | 3 | 490 |

More Information
Other Species Targeted

Gene Ontology Categories:
