siRNA Details
Gene Symbol: Igf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001111274.1 | NP_001104744.1 | 4 | 627 | ||
NM_010512.4 | NP_034642.2 | 4 | 709 | |||
NM_184052.3 | NP_908941.1 | 4 | 627 | |||
GenBank mRNA | AK038119.1 | - | 4 | 1710 | ||
AY878193.1 | - | 4 | 507 | |||
BC012409.1 | - | 3 | 364 | 4194295 | ||
CT010364.1 | - | 3 | 337 | |||
X04482.1 | - | 4 | 409 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
