siRNA Details
Gene Symbol: Igf2r | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010515.2 | NP_034645.2 | 42 | 6423 | ||
GenBank mRNA | AK053045.1 | - | 7 | 1015 | ||
AK088699.1 | - | 17 | 2649 | |||
BC031190.1 | - | 9 | 1429 | |||
BC043457.1 | - | 5 | 624 | |||
BC055018.1 | - | 5 | 629 | |||
BC150817.1 | - | 42 | 6334 | 9087728 | ||
BC171973.1 | - | 42 | 6334 | 9054259 | ||
U04710.1 | - | 42 | 6379 |
