siRNA Details
Gene Symbol: Igfbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008341.4 | NP_032367.3 | 3 | 757 | ||
GenBank mRNA | BC013345.1 | - | 3 | 788 | 4161889 | |
X81579.1 | - | 3 | 677 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
