siRNA Details
Gene Symbol: Igfbp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008343.2 | NP_032369.2 | 2 | 698 | ||
GenBank mRNA | AK077477.1 | - | 2 | 239 | ||
AK141690.1 | - | 2 | 320 | |||
BC031421.1 | - | 2 | 657 | |||
BC058261.1 | - | 2 | 668 | 6487655 | ||
X81581.1 | - | 2 | 646 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
