siRNA Details
Gene Symbol: Igfbp5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010518.2 | NP_034648.2 | 3 | 1382 | ||
GenBank mRNA | AK142108.1 | - | 3 | 1402 | ||
AK161584.1 | - | 2 | 191 | |||
BC026805.1 | - | 3 | 822 | |||
BC029321.1 | - | 2 | 216 | |||
BC054812.1 | - | 3 | 792 | 6841070 | ||
BC057447.1 | - | 3 | 1382 | 2648602 | ||
L12447.1 | - | 3 | 1043 | |||
X81583.1 | - | 3 | 719 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
