siRNA Details
Gene Symbol: Igh-V15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | U39293.1 | - | 2 | 152 | ||
U39294.1 | - | 2 | 152 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Igh-V15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | U39293.1 | - | 2 | 152 | ||
U39294.1 | - | 2 | 152 |