siRNA Details
Gene Symbol: Ighv7-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | M12422.1 | - | 1 | 12 | ||
M58659.1 | - | 167 |
Gene Symbol: Igh-VJ558 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF144975.1 | - | 1 | 122 |
Gene Symbol: Igh-VS107 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF045498.1 | - | 2 | 224 | ||
BC111444.1 | - | 1 | 494 | |||
M16334.1 | - | 2 | 269 | |||
M37672.1 | - | 1 | 167 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
