siRNA Details
Gene Symbol: Il18bp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010531.1 | NP_034661.1 | 7 | 1072 | ||
GenBank mRNA | AF110802.1 | - | 4 | 2506 | ||
AF122907.1 | - | 7 | 1072 | |||
AK003370.1 | - | 7 | 1088 | |||
AK008452.1 | - | 6 | 1003 | |||
AK009721.1 | - | 6 | 999 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
