siRNA Details
Gene Symbol: Il18bp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010531.1 | NP_034661.1 | 4 | 311 | ||
GenBank mRNA | AB019505.1 | - | 2 | 160 | ||
AF110802.1 | - | 2 | 1538 | |||
AF110803.1 | - | 3 | 238 | |||
AF122907.1 | - | 4 | 311 | |||
AK003370.1 | - | 4 | 327 | |||
AK008452.1 | - | 3 | 242 | |||
AK009721.1 | - | 3 | 238 | |||
BC018332.1 | - | 3 | 221 | 4007411 | ||
CT010164.1 | - | 2 | 160 | |||
CT010334.1 | - | 2 | 160 |

More Information
Other Species Targeted

Gene Ontology Categories:
