siRNA Details
Gene Symbol: Il10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010548.2 | NP_034678.1 | 1, 2 | 225 | ||
GenBank mRNA | AK152344.1 | - | 1, 2 | 226 | ||
BC120612.1 | - | 1, 2 | 200 | 40129535 | ||
BC137844.1 | - | 1, 2 | 200 | 8860863 | ||
M37897.1 | - | 1, 2 | 233 |
