siRNA Details
Gene Symbol: Il18r1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161842.1 | NP_001155314.1 | 6 | 1031 | ||
NM_001161843.1 | NP_001155315.1 | 6 | 903 | |||
NM_008365.2 | NP_032391.1 | 5 | 956 | |||
GenBank mRNA | AK089237.1 | - | 6 | 1032 | ||
AK170705.1 | - | 6 | 992 | |||
BC023240.1 | - | 6 | 872 | 4483261 | ||
U43673.1 | - | 5 | 956 |
