siRNA Details
Gene Symbol: Il6st | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010560.3 | NP_034690.3 | 10 | 1441 | ||
GenBank mRNA | BC020454.1 | - | 10 | 1395 | ||
BC058679.1 | - | 10 | 1402 | 6834623 | ||
M83336.1 | - | 9 | 1246 | |||
X62646.1 | - | 9 | 1246 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
