siRNA Details
Gene Symbol: Incenp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016692.3 | NP_057901.2 | 4 | 522 | ||
GenBank mRNA | AB100432.1 | - | 3 | 316 | ||
AB100433.1 | - | 3 | 316 | |||
AF117610.1 | - | 4 | 458 | |||
AK034764.1 | - | 4 | 451 | |||
AK081841.1 | - | 4 | 456 | |||
AK088627.1 | - | 4 | 479 | |||
BC021761.2 | - | 4 | 419 | |||
BC037011.1 | - | 4 | 475 | |||
BC052414.1 | - | 4 | 455 | 6401188 |
