siRNA Details
Gene Symbol: Inha | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010564.4 | NP_034694.3 | 2 | 1216 | ||
GenBank mRNA | BC056627.1 | - | 2 | 1216 | 5009577 | |
X55957.1 | - | 2 | 902 | |||
X69618.1 | - | 2 | 1068 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
