siRNA Details
Gene Symbol: Irf4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013674.1 | NP_038702.1 | 9 | 1362 | ||
GenBank mRNA | AK089319.1 | - | 9 | 1381 | ||
AK089819.1 | - | 9 | 1384 | |||
AK154138.1 | - | 9 | 1386 | |||
AK154315.1 | - | 9 | 1374 | |||
AK156562.1 | - | 9 | 1383 | |||
BC137713.1 | - | 9 | 1350 | 8860732 | ||
BC137714.1 | - | 9 | 1350 | 8860733 | ||
U11692.1 | - | 9 | 1362 | |||
U34307.1 | - | 9 | 1356 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
