siRNA Details
Gene Symbol: Itga7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008398.2 | NP_032424.2 | 17 | 2484 | ||
GenBank mRNA | AK157612.1 | - | 17 | 2491 | ||
CX208029.1 | - | 4 | 350 | |||
L23423.1 | - | 17 | 2482 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
