siRNA Details
Gene Symbol: Itgav | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008402.3 | NP_032428.2 | 2 | 640 | ||
GenBank mRNA | AK140647.1 | - | 2 | 582 | ||
AK149984.1 | - | 2 | 542 | |||
BB632484.1 | - | 2 | 637 | |||
BC133682.1 | - | 2 | 352 | |||
U14135.1 | - | 2 | 522 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
