siRNA Details
Gene Symbol: Itgav | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008402.3 | NP_032428.2 | 15 | 1772 | ||
GenBank mRNA | AK149984.1 | - | 15 | 1673 | ||
AK154307.1 | - | 3 | 168 | |||
BC133682.1 | - | 15 | 1483 | |||
U14135.1 | - | 15 | 1654 |

More Information
Other Species Targeted

Gene Ontology Categories:
