siRNA Details
Gene Symbol: Itgav | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008402.3 | NP_032428.2 | 28, 29 | 3255 | ||
GenBank mRNA | AK154307.1 | - | 16, 17 | 1651 | ||
BC048857.1 | - | 3, 4 | 312 | |||
BC133682.1 | - | 28, 29 | 2966 | |||
CN532926.1 | - | 3, 4 | 320 | |||
U14135.1 | - | 28, 29 | 3137 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
