siRNA Details
Gene Symbol: Itgb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016780.2 | NP_058060.2 | 4 | 601 | ||
GenBank mRNA | AF026509.1 | - | 4 | 545 | ||
AK157958.1 | - | 4 | 547 | |||
AY541673.1 | - | 4 | 515 | |||
BC125518.1 | - | 4 | 601 | 40130033 | ||
BC125520.1 | - | 4 | 601 | 40130035 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
