siRNA Details
Gene Symbol: Itih3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008407.2 | NP_032433.2 | 18 | 2037 | ||
GenBank mRNA | AK146700.1 | - | 17 | 2526 | ||
BC015276.1 | - | 18 | 1995 | 4212290 | ||
X70393.1 | - | 17 | 1994 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
