siRNA Details
Gene Symbol: Itm2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008409.2 | NP_032435.2 | 5 | 816 | ||
GenBank mRNA | AK014024.1 | - | 5 | 584 | ||
AK051956.1 | - | 5 | 795 | |||
AK135229.1 | - | 5 | 768 | |||
AK167351.1 | - | 5 | 817 | |||
BC031420.1 | - | 5 | 783 | 3668557 | ||
L38971.1 | - | 5 | 819 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
