siRNA Details
Gene Symbol: Itm2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008410.2 | NP_032436.1 | 4 | 614 | ||
GenBank mRNA | AB030203.1 | - | 4 | 634 | ||
AB030204.1 | - | 3 | 430 | |||
AK002599.1 | - | 4 | 577 | |||
AK003224.1 | - | 4 | 581 | |||
AK003587.1 | - | 4 | 603 | |||
AK003928.1 | - | 4 | 602 | |||
AK005125.1 | - | 4 | 605 | |||
AK076139.1 | - | 4 | 623 | |||
AK077482.1 | - | 4 | 616 | |||
AK088270.1 | - | 4 | 601 | |||
AK150378.1 | - | 4 | 634 | |||
AK150396.1 | - | 4 | 633 | |||
AK151714.1 | - | 4 | 633 | |||
AK151889.1 | - | 4 | 634 | |||
AK152068.1 | - | 4 | 622 | |||
AK152516.1 | - | 4 | 634 | |||
AK152650.1 | - | 4 | 634 | |||
AK152731.1 | - | 4 | 605 | |||
AK152983.1 | - | 4 | 626 | |||
AK153103.1 | - | 4 | 577 | |||
AK159307.1 | - | 4 | 577 | |||
AK159340.1 | - | 4 | 602 | |||
AK159378.1 | - | 4 | 633 | |||
AK159426.1 | - | 4 | 602 | |||
AK159619.1 | - | 4 | 626 | |||
AK159628.1 | - | 4 | 602 | |||
AK159792.1 | - | 4 | 602 | |||
AK159962.1 | - | 4 | 605 | |||
AK160101.1 | - | 4 | 602 | |||
AK160725.1 | - | 4 | 633 | |||
AK194069.1 | - | 179 | ||||
BC004731.1 | - | 4 | 585 | 3583154 | ||
BC010320.1 | - | 4 | 617 | 3599190 | ||
BC021786.1 | - | 4 | 623 | 4458918 | ||
U76253.1 | - | 4 | 614 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
