siRNA Details
Gene Symbol: Itsn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110275.1 | NP_001103745.1 | 26 | 3488 | ||
NM_010587.2 | NP_034717.2 | 26 | 3488 | |||
GenBank mRNA | AF132478.1 | - | 25 | 3227 | ||
AF132481.1 | - | 25 | 3227 | |||
AF169621.1 | - | 5 | 334 | |||
AF498310.1 | - | 4 | 167 | |||
AF498311.1 | - | 3 | 121 | |||
AK082606.1 | - | 5 | 351 | |||
AK160794.1 | - | 11 | 1477 | |||
AK161183.1 | - | 1 | 1249 | |||
BC062938.1 | - | 26 | 3487 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
