siRNA Details
Gene Symbol: Itsn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110275.1 | NP_001103745.1 | 13 | 1598 | ||
NM_001110276.1 | NP_001103746.1 | 13 | 1598 | |||
NM_010587.2 | NP_034717.2 | 13 | 1598 | |||
GenBank mRNA | AF132478.1 | - | 12 | 1337 | ||
AF132481.1 | - | 12 | 1337 | |||
AK155397.1 | - | 13 | 1553 | |||
AK160949.1 | - | 13 | 1599 | |||
BC062938.1 | - | 13 | 1598 | |||
BC066105.1 | - | 13 | 1460 |
