siRNA Details
Gene Symbol: Itsn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010587.2 | NP_034717.2 | 32 | 4399 | ||
GenBank mRNA | AF132481.1 | - | 31 | 4138 | ||
AK082606.1 | - | 11 | 1262 | |||
AY257970.1 | - | 4 | 452 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
