siRNA Details
Gene Symbol: Jag1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013822.5 | NP_038850.1 | 20 | 2669 | ||
GenBank mRNA | AF171092.1 | - | 20 | 2674 | ||
AK137092.1 | - | 20 | 2569 | |||
AK162228.1 | - | 3 | 171 | |||
BC058675.1 | - | 20 | 2481 | 6834418 |
