siRNA Details
Gene Symbol: Jun | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010591.2 | NP_034721.1 | 1 | 1062 | ||
GenBank mRNA | AK132019.1 | - | 1 | 1037 | ||
AK133722.1 | - | 1 | 1042 | |||
AK133867.1 | - | 1 | 1044 | |||
AK135023.1 | - | 1 | 1041 | |||
AK140911.1 | - | 1 | 1037 | |||
AK159196.1 | - | 1 | 1037 | |||
AK159220.1 | - | 1 | 1042 | |||
BC002081.1 | - | 1 | 205 | 3493248 | ||
BC021888.1 | - | 1 | 996 | 3156091 | ||
BC032086.1 | - | 1 | 530 | |||
BC055887.1 | - | 1 | 1139 | |||
BC094032.1 | - | 2 | 924 | 4936392 | ||
J04115.1 | - | 1 | 1027 | |||
X12740.1 | - | 1 | 725 | |||
X12761.1 | - | 1 | 465 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
