siRNA Details
Gene Symbol: Jup | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010593.2 | NP_034723.1 | 12 | 2232 | ||
GenBank mRNA | AK086401.1 | - | 1 | 48 | ||
AK145895.1 | - | 12 | 2217 | |||
AK170934.1 | - | 12 | 2205 | |||
BC040757.1 | - | 12 | 2181 | 4216772 | ||
BC094461.1 | - | 12 | 2185 | 4459882 | ||
M90365.1 | - | 10 | 1697 |

More Information
Other Species Targeted

Gene Ontology Categories:
