siRNA Details
Gene Symbol: Kap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010594.2 | NP_034724.1 | 1, 2 | 114 | ||
GenBank mRNA | AK142506.1 | - | 1, 2 | 115 | ||
AV000934.1 | - | 1, 2 | 131 | |||
BC013453.1 | - | 1, 2 | 100 | 4221610 | ||
M22810.1 | - | 1, 2 | 113 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
