siRNA Details
Gene Symbol: Kcna1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010595.3 | NP_034725.3 | 2 | 3476 | ||
GenBank mRNA | AK039559.1 | - | 2 | 2471 | ||
BC112970.2 | - | 2 | 2081 | 40048563 | ||
Y00305.1 | - | 1 | 2855 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
