siRNA Details
Gene Symbol: Kcne1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008424.3 | NP_032450.1 | 2 | 399 | ||
GenBank mRNA | AK008938.1 | - | 2 | 394 | ||
AK028907.1 | - | 2 | 400 | |||
BC094276.1 | - | 2 | 346 | 4189679 | ||
X60457.1 | - | 2 | 276 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
