siRNA Details
Gene Symbol: Kcnk7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010609.2 | NP_034739.2 | 3 | 829 | ||
GenBank mRNA | AB015729.1 | - | 3 | 874 | ||
AF012324.1 | - | 3 | 889 | |||
AF022820.1 | - | 3 | 826 | |||
AF110521.1 | - | 3 | 940 | |||
AK160180.1 | - | 3 | 836 | |||
BC106993.2 | - | 3 | 829 | 40044182 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
