siRNA Details
Gene Symbol: Kcnq1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008434.2 | NP_032460.2 | 10 | 1417 | ||
GenBank mRNA | AB079603.1 | - | 10 | 1120 | ||
AK136389.1 | - | 3 | 287 | |||
AK154244.1 | - | 10 | 1391 | |||
AY331142.1 | - | 10 | 1332 | |||
BC045142.1 | - | 10 | 1417 | 6314642 | ||
BC055304.1 | - | 10 | 1372 | 6397978 | ||
U70068.1 | - | 10 | 1223 |
