siRNA Details
Gene Symbol: Kcnq2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB000494.1 | - | 3 | 506 | ||
AB000495.1 | - | 3 | 506 | |||
AB000496.1 | - | 3 | 506 | |||
AB000497.1 | - | 3 | 506 | |||
AB000498.1 | - | 3 | 506 | |||
AB000499.1 | - | 3 | 506 | |||
AB000500.1 | - | 3 | 506 | |||
AB000501.1 | - | 3 | 506 | |||
AB000502.1 | - | 3 | 506 | |||
AB000503.1 | - | 3 | 506 | |||
AB000504.1 | - | 3 | 506 | |||
AF490773.1 | - | 3 | 420 | |||
AK135059.1 | - | 2 | 78 | |||
AK139411.1 | - | 3 | 251 | |||
AK161558.1 | - | 2 | 1512 | |||
BC145453.1 | - | 3 | 550 | 9054011 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
