siRNA Details
Gene Symbol: Kif16b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081133.2 | NP_001074602.1 | 26 | 4984 | ||
GenBank mRNA | AB610968.1 | - | 27 | 5072 | ||
AK018455.1 | - | 2 | 1454 | |||
AK129401.1 | - | 14 | 3188 | |||
AK142909.1 | - | 8 | 2285 | |||
AK160835.1 | - | 10 | 3204 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
