siRNA Details
Gene Symbol: Kif21a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109040.2 | NP_001102510.1 | 10 | 1669 | ||
NM_001109041.2 | NP_001102511.1 | 10 | 1669 | |||
NM_001109042.2 | NP_001102512.1 | 10 | 1669 | |||
NM_016705.4 | NP_057914.2 | 10 | 1669 | |||
GenBank mRNA | AF202892.1 | - | 10 | 1529 | ||
AK019525.1 | - | 10 | 1670 | |||
AK047350.1 | - | 10 | 1559 | |||
AK047880.1 | - | 10 | 1558 | |||
AK129426.1 | - | 3 | 277 | |||
AK147436.1 | - | 10 | 1557 | |||
AK163536.1 | - | 10 | 1553 | |||
BC060698.1 | - | 10 | 1536 | 6409819 | ||
BC062896.1 | - | 10 | 1536 | 6409819 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
