siRNA Details
Gene Symbol: Klf9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010638.4 | NP_034768.2 | 1 | 1000 | ||
GenBank mRNA | AK028544.1 | - | 1 | 1003 | ||
BC106117.1 | - | 2 | 827 | |||
BC138724.1 | - | 1 | 818 | 8861746 | ||
BC138725.1 | - | 1 | 813 | 8861747 | ||
CT010185.1 | - | 1 | 471 | |||
Y14296.1 | - | 1 | 471 |

More Information
Other Species Targeted

Gene Ontology Categories:
